Assay Details
Gene Symbol: PAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095275.4 | XP_001095275.3 | 1051 | 88 | ||
XM_015149591.2 | XP_015005077.1 | 1051 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
