Assay Details
Gene Symbol: VRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112417.2 | XP_014967903.2 | 458 | 148 | ||
XM_028832465.1 | XP_028688298.1 | 458 | 148 | |||
XM_028832466.1 | XP_028688299.1 | 458 | 148 | |||
XM_028832467.1 | XP_028688300.1 | 458 | 148 | |||
XM_028832468.1 | XP_028688301.1 | 595 | 148 | |||
GenBank mRNA | JU336249.1 | - | 378 | 148 | ||
JU336250.1 | - | 378 | 148 | |||
JU348056.1 | - | 378 | 148 |
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU336251.1 | - | 378 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028832468.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
