Assay Details
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109499.4 | XP_001109499.1 | 1209 | 61 | ||
GenBank mRNA | JU325904.1 | - | 620 | 61 | ||
JU470475.1 | - | 917 | 61 | |||
JV044169.1 | - | 913 | 61 | |||
JV635420.1 | - | 923 | 61 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
