Assay Details
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190973.2 | NP_001177902.1 | 2 - 3 | 292 | 111 | |
GenBank mRNA | JU320757.1 | - | 2 - 3 | 291 | 111 | |
JU471088.1 | - | 2 - 3 | 297 | 111 | ||
JV044525.1 | - | 2 - 3 | 295 | 111 | ||
JV491241.1 | - | 2 - 3 | 211 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
