Assay Details
Gene Symbol: LOC717978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120768.2 | XP_014976254.2 | 589 | 152 | ||
GenBank mRNA | JU333035.1 | - | 5 - 6 | 586 | 152 | |
JU333036.1 | - | 5 - 6 | 586 | 152 |
Gene Symbol: PARP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028837587.1 | XP_028693420.1 | 582 | 152 | ||
GenBank mRNA | JV046242.1 | - | 5 - 6 | 554 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() |
