Assay Details
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193910.1 | NP_001180839.1 | 2 - 3 | 262 | 103 | |
GenBank mRNA | JV043793.1 | - | 2 - 3 | 265 | 103 | |
JV638916.1 | - | 2 - 3 | 262 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
