Assay Details
Gene Symbol: KDELR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261069.1 | NP_001247998.1 | 536 | 114 | ||
XM_028838489.1 | XP_028694322.1 | 354 | 114 | |||
GenBank mRNA | JU332650.1 | - | 538 | 114 | ||
JU474093.1 | - | 536 | 114 | |||
JV492778.1 | - | 388 | 114 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
