Assay Details
Gene Symbol: SLC19A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257523.1 | NP_001244452.1 | 870 | 151 | ||
XM_028835438.1 | XP_028691271.1 | 1232 | 151 | |||
GenBank mRNA | JU332537.1 | - | 870 | 151 | ||
JU474013.1 | - | 850 | 151 | |||
JV045940.1 | - | 1007 | 151 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
