Assay Details
Gene Symbol: ZNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127294.2 | XP_014982780.1 | 396 | 116 | ||
GenBank mRNA | JV636363.1 | - | 1 - 2 | 105 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
