Assay Details
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266170.1 | NP_001253099.1 | 416 | 107 | ||
GenBank mRNA | JU320145.1 | - | 340 | 107 | ||
JV044217.1 | - | 416 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
