Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260566.1 | NP_001247495.1 | 10 - 11 | 1793 | 80 | |
GenBank mRNA | JU333443.1 | - | 10 - 11 | 1809 | 80 | |
JV046462.1 | - | 10 - 11 | 1793 | 80 | ||
JV609123.1 | - | 10 - 11 | 1816 | 80 | ||
JV636033.1 | - | 10 - 11 | 1799 | 80 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
