Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260566.1 | NP_001247495.1 | 12 - 13 | 2135 | 157 | |
XM_015119568.2 | XP_014975054.1 | 3249 | 157 | |||
GenBank mRNA | JU333443.1 | - | 12 - 13 | 2151 | 157 | |
JV046462.1 | - | 12 - 13 | 2135 | 157 | ||
JV609123.1 | - | 12 - 13 | 2158 | 157 | ||
JV636033.1 | - | 12 - 13 | 2141 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
