Assay Details
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095528.3 | XP_001095528.1 | 246 | 123 | ||
GenBank mRNA | JU484345.1 | - | 1 - 2 | 120 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
