Assay Details
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261365.1 | NP_001248294.1 | 242 | 93 | ||
GenBank mRNA | JU323181.1 | - | 233 | 93 | ||
JU473317.1 | - | 244 | 93 | |||
JV045278.1 | - | 242 | 93 |
