Assay Details
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261365.1 | NP_001248294.1 | 723 | 85 | ||
GenBank mRNA | JU323181.1 | - | 714 | 85 | ||
JU473317.1 | - | 725 | 85 | |||
JV045278.1 | - | 723 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
