Assay Details
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261123.1 | NP_001248052.1 | 2 - 3 | 186 | 100 | |
XM_015115282.2 | XP_014970768.1 | 410 | 100 | |||
XM_015115284.2 | XP_014970770.1 | 799 | 100 | |||
XM_015115287.2 | XP_014970773.1 | 637 | 100 | |||
XM_015115289.2 | XP_014970775.1 | 451 | 100 | |||
XM_015115290.2 | XP_014970776.1 | 575 | 100 | |||
XM_028832593.1 | XP_028688426.1 | 1073 | 100 | |||
XM_028832594.1 | XP_028688427.1 | 893 | 100 | |||
XM_028832595.1 | XP_028688428.1 | 893 | 100 | |||
GenBank mRNA | JU334030.1 | - | 3 - 4 | 284 | 100 | |
JV046779.1 | - | 2 - 3 | 186 | 100 | ||
JV648511.1 | - | 2 - 3 | 193 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
