Assay Details
Gene Symbol: SAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU476716.1 | - | 1454 | 92 |
Gene Symbol: SAFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084042.3 | XP_001084042.2 | 1734 | 92 | ||
GenBank mRNA | JU324059.1 | - | 10 - 11 | 1522 | 92 | |
JV076380.1 | - | 10 - 11 | 1552 | 92 | ||
JV727623.1 | - | 1507 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
