Assay Details
Gene Symbol: SAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU476716.1 | - | 1300 | 90 |
Gene Symbol: SAFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084042.3 | XP_001084042.2 | 1580 | 90 | ||
GenBank mRNA | JU324059.1 | - | 9 - 10 | 1368 | 90 | |
JV076380.1 | - | 9 - 10 | 1398 | 90 | ||
JV727623.1 | - | 1353 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
