Assay Details
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122249.2 | XP_014977735.1 | 858 | 149 | ||
XM_015122250.2 | XP_014977736.1 | 645 | 149 | |||
XM_015122251.2 | XP_014977737.1 | 287 | 149 | |||
XM_015122252.2 | XP_014977738.1 | 259 | 149 | |||
XM_028837904.1 | XP_028693737.1 | 1190 | 149 | |||
XM_028837905.1 | XP_028693738.1 | 1070 | 149 | |||
GenBank mRNA | JU324312.1 | - | 3 - 4 | 317 | 149 | |
JU474716.1 | - | 2 - 3 | 216 | 149 | ||
JV709779.1 | - | 3 - 4 | 309 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
