Assay Details
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136529.2 | XP_014992015.1 | 2049 | 96 | ||
XM_015136532.2 | XP_014992018.1 | 2027 | 96 | |||
XM_028847479.1 | XP_028703312.1 | 5473 | 96 | |||
XM_028847480.1 | XP_028703313.1 | 2188 | 96 | |||
XM_028847481.1 | XP_028703314.1 | 5451 | 96 | |||
GenBank mRNA | JU321188.1 | - | 1999 | 96 | ||
JU321189.1 | - | 1999 | 96 | |||
JU474719.1 | - | 1989 | 96 | |||
JU474720.1 | - | 1989 | 96 | |||
JU474721.1 | - | 1989 | 96 | |||
JU474722.1 | - | 1989 | 96 | |||
JU474723.1 | - | 1989 | 96 | |||
JU474724.1 | - | 1989 | 96 | |||
JU474725.1 | - | 1989 | 96 | |||
JU474726.1 | - | 1989 | 96 | |||
JV046466.1 | - | 1999 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
