Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083373.4 | XP_001083373.1 | 1301 | 142 | ||
XM_001083494.4 | XP_001083494.1 | 1204 | 142 | |||
XM_015120917.2 | XP_014976403.1 | 1176 | 142 | |||
XM_015120918.2 | XP_014976404.1 | 1275 | 142 | |||
XM_015120919.2 | XP_014976405.1 | 1141 | 142 | |||
GenBank mRNA | JU333558.1 | - | 3 - 4 | 1099 | 142 | |
JV710201.1 | - | 3 - 4 | 1093 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() |
