Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083373.4 | XP_001083373.1 | 1939 | 96 | ||
XM_001083494.4 | XP_001083494.1 | 1842 | 96 | |||
XM_015120917.2 | XP_014976403.1 | 1814 | 96 | |||
XM_015120918.2 | XP_014976404.1 | 1913 | 96 | |||
XM_015120919.2 | XP_014976405.1 | 1779 | 96 | |||
GenBank mRNA | JV710201.1 | - | 7 - 8 | 1731 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
