Assay Details
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126683.2 | XP_014982169.2 | 679 | 162 | ||
XM_015126684.2 | XP_014982170.2 | 1346 | 162 | |||
XM_015126685.2 | XP_014982171.2 | 1323 | 162 | |||
XM_015126686.2 | XP_014982172.2 | 401 | 162 | |||
GenBank mRNA | JU326208.1 | - | 428 | 162 | ||
JU326209.1 | - | 428 | 162 | |||
JV646539.1 | - | 496 | 162 | |||
JV646540.1 | - | 496 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() |
