Assay Details
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266634.1 | NP_001253563.1 | 1 - 2 | 141 | 133 | |
XM_015134313.2 | XP_014989799.1 | 278 | 133 | |||
XM_028844949.1 | XP_028700782.1 | 271 | 133 | |||
GenBank mRNA | JU333845.1 | - | 1 - 2 | 141 | 133 | |
JV602605.1 | - | 1 - 2 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
