Assay Details
Gene Symbol: PNKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028831102.1 | XP_028686935.1 | 520 | 98 | ||
XM_028831103.1 | XP_028686936.1 | 576 | 98 | |||
GenBank mRNA | JU327047.1 | - | 3 - 4 | 385 | 98 | |
JV057351.1 | - | 3 - 4 | 389 | 98 | ||
JV463925.1 | - | 4 - 5 | 423 | 98 | ||
JV540369.1 | - | 4 - 5 | 463 | 98 | ||
JV600267.1 | - | 3 - 4 | 474 | 98 | ||
JV647749.1 | - | 2 - 3 | 217 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
