Assay Details
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260536.1 | NP_001247465.1 | 3 - 4 | 818 | 88 | |
XM_015114182.2 | XP_014969668.1 | 1497 | 88 | |||
XM_015114183.2 | XP_014969669.1 | 910 | 88 | |||
XM_015114184.2 | XP_014969670.1 | 986 | 88 | |||
GenBank mRNA | JV047607.1 | - | 3 - 4 | 818 | 88 |
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV706336.1 | - | 4 - 5 | 2845 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
