Assay Details
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257429.1 | NP_001244358.1 | 480 | 145 | ||
XM_015124569.2 | XP_014980055.1 | 718 | 145 | |||
XM_015124577.2 | XP_014980063.1 | 513 | 145 | |||
XM_015124586.2 | XP_014980072.1 | 760 | 145 | |||
XM_028829820.1 | XP_028685653.1 | 620 | 145 | |||
XM_028829848.1 | XP_028685681.1 | 691 | 145 | |||
GenBank mRNA | JU335661.1 | - | 480 | 145 | ||
JV044794.1 | - | 481 | 145 | |||
JV567514.1 | - | 470 | 145 | |||
JV601143.1 | - | 481 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
