Assay Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091031.4 | XP_001091031.3 | 1122 | 178 | ||
XM_015132987.2 | XP_014988473.2 | 1123 | 178 |

More Information
Set Membership: |
![]() ![]() |
