Assay Details
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260688.1 | NP_001247617.1 | 163 | 81 | ||
XM_015123306.2 | XP_014978792.1 | 191 | 81 | |||
XM_015123309.2 | XP_014978795.1 | 191 | 81 | |||
XM_028838551.1 | XP_028694384.1 | 191 | 81 | |||
GenBank mRNA | JU473664.1 | - | 163 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
