Assay Details
Gene Symbol: ADPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110060.4 | XP_001110060.2 | 782 | 130 | ||
GenBank mRNA | JV636890.1 | - | 4 - 5 | 729 | 130 | |
JV704694.1 | - | 4 - 5 | 707 | 130 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
