Assay Details
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260739.1 | NP_001247668.1 | 646 | 128 | ||
GenBank mRNA | JU320685.1 | - | 632 | 128 | ||
JU472487.1 | - | 667 | 128 | |||
JV047478.1 | - | 646 | 128 | |||
JV654843.1 | - | 671 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
