Assay Details
Gene Symbol: RADX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193976.1 | NP_001180905.1 | 10 - 11 | 2054 | 138 | |
XM_028841737.1 | XP_028697570.1 | 1763 | 138 | |||
GenBank mRNA | JU335224.1 | - | 10 - 11 | 2049 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
