Assay Details
Gene Symbol: RADX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193976.1 | NP_001180905.1 | 12 - 13 | 2423 | 92 | |
XM_028841737.1 | XP_028697570.1 | 2132 | 92 | |||
GenBank mRNA | JU335224.1 | - | 12 - 13 | 2418 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
