Assay Details
Gene Symbol: RADX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193976.1 | NP_001180905.1 | 7 - 8 | 1559 | 132 | |
GenBank mRNA | JU335224.1 | - | 7 - 8 | 1554 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
