Assay Details
Gene Symbol: RADX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193976.1 | NP_001180905.1 | 8 - 9 | 1726 | 97 | |
GenBank mRNA | JU335224.1 | - | 8 - 9 | 1721 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
