Assay Details
Gene Symbol: DYNC2I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846530.1 | XP_028702363.1 | 2514 | 177 | ||
XM_028846531.1 | XP_028702364.1 | 2472 | 177 | |||
XM_028846532.1 | XP_028702365.1 | 2372 | 177 | |||
XM_028846533.1 | XP_028702366.1 | 2514 | 177 | |||
XM_028846534.1 | XP_028702367.1 | 2416 | 177 | |||
XM_028846535.1 | XP_028702368.1 | 1918 | 177 | |||
XM_028846536.1 | XP_028702369.1 | 1341 | 177 | |||
XM_028846537.1 | XP_028702370.1 | 1860 | 177 | |||
XM_028846538.1 | XP_028702371.1 | 1762 | 177 | |||
XM_028846539.1 | XP_028702372.1 | 1654 | 177 | |||
XM_028846540.1 | XP_028702373.1 | 1794 | 177 |

More Information
Set Membership: |
![]() ![]() |
