Assay Details
Gene Symbol: SMG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194385.1 | NP_001181314.1 | 2811 | 116 | ||
GenBank mRNA | JU335254.1 | - | 2769 | 116 | ||
JU489100.1 | - | 2757 | 116 | |||
JV586108.1 | - | 1880 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
