Assay Details
Gene Symbol: LRIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266807.1 | NP_001253736.1 | 1831 | 128 | ||
XM_015146984.2 | XP_015002470.2 | 1825 | 128 | |||
GenBank mRNA | JU476379.1 | - | 1831 | 128 | ||
JV047624.1 | - | 1831 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
