Assay Details
Gene Symbol: PXMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261587.2 | NP_001248516.1 | 4 - 5 | 535 | 146 | |
GenBank mRNA | JU322989.1 | - | 4 - 5 | 540 | 146 | |
JV044129.1 | - | 4 - 5 | 535 | 146 | ||
JV668702.1 | - | 4 - 5 | 589 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
