Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104586.4 | XP_001104586.2 | 141 | 86 | ||
XM_015116481.2 | XP_014971967.1 | 142 | 86 | |||
GenBank mRNA | JU323248.1 | - | 97 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001104586.4, XM_015116481.2 transcripts.
Set Membership: |
![]() ![]() |
