Assay Details
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265767.1 | NP_001252696.1 | 461 | 77 | ||
GenBank mRNA | JU484540.1 | - | 354 | 77 | ||
JV635765.1 | - | 461 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
