Assay Details
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261810.1 | NP_001248739.1 | 1 - 2 | 241 | 120 | |
GenBank mRNA | JU321789.1 | - | 1 - 2 | 240 | 120 | |
JU473370.1 | - | 1 - 2 | 241 | 120 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
