Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133060.2 | XP_014988546.1 | 605 | 103 | ||
XM_015133062.2 | XP_014988548.1 | 429 | 103 | |||
XM_028845385.1 | XP_028701218.1 | 206 | 103 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015133062.2, XM_028845385.1 transcripts.
This assay is in the 5'UTR region of the XM_015133062.2, XM_028845385.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
