Assay Details
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028830088.1 | XP_028685921.1 | 932 | 136 | ||
XM_028830089.1 | XP_028685922.1 | 932 | 136 | |||
XM_028830090.1 | XP_028685923.1 | 428 | 136 | |||
XM_028830091.1 | XP_028685924.1 | 810 | 136 | |||
XM_028830092.1 | XP_028685925.1 | 635 | 136 | |||
GenBank mRNA | JU326980.1 | - | 449 | 136 | ||
JU326981.1 | - | 449 | 136 | |||
JU326982.1 | - | 449 | 136 | |||
JU486277.1 | - | 502 | 136 | |||
JU486278.1 | - | 502 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
