Assay Details
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144493.2 | XP_014999979.1 | 382 | 59 | ||
XM_028851835.1 | XP_028707668.1 | 450 | 59 | |||
XM_028851836.1 | XP_028707669.1 | 328 | 59 | |||
XM_028851837.1 | XP_028707670.1 | 367 | 59 | |||
XM_028851838.1 | XP_028707671.1 | 503 | 59 | |||
XM_028851839.1 | XP_028707672.1 | 450 | 59 | |||
XM_028851840.1 | XP_028707673.1 | 404 | 59 | |||
GenBank mRNA | JV623787.1 | - | 3 - 4 | 254 | 59 | |
JV667433.1 | - | 3 - 4 | 256 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
