Assay Details
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042433.2 | NP_001035898.1 | 1470 | 117 | ||
XM_015121466.2 | XP_014976952.1 | 1744 | 117 | |||
GenBank mRNA | DQ644021.1 | - | 1473 | 117 | ||
EF467993.1 | - | 1472 | 117 | |||
JU470652.1 | - | 1641 | 117 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
