Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116117.2 | XP_014971603.2 | 287 | 135 | ||
XM_015116118.2 | XP_014971604.2 | 380 | 135 | |||
GenBank mRNA | JU476945.1 | - | 2 - 3 | 263 | 135 | |
JV069574.1 | - | 2 - 3 | 241 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
