Assay Details
Gene Symbol: HPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149965.2 | XP_015005451.2 | 936 | 91 | ||
XM_015149966.2 | XP_015005452.2 | 845 | 91 | |||
XM_015149967.2 | XP_015005453.2 | 936 | 91 | |||
XM_028827904.1 | XP_028683737.1 | 928 | 91 | |||
XM_028827905.1 | XP_028683738.1 | 845 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
