Assay Details
Gene Symbol: MTHFSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085469.4 | XP_001085469.4 | 494 | 152 | ||
XM_001085714.4 | XP_001085714.4 | 502 | 152 | |||
XM_015126738.2 | XP_014982224.1 | 665 | 152 | |||
XM_015126739.2 | XP_014982225.1 | 667 | 152 | |||
XM_015126740.2 | XP_014982226.2 | 502 | 152 | |||
XM_015126741.2 | XP_014982227.2 | 494 | 152 | |||
XM_015126742.2 | XP_014982228.1 | 545 | 152 | |||
XM_015126743.2 | XP_014982229.1 | 548 | 152 | |||
XM_015126745.2 | XP_014982231.1 | 1969 | 152 | |||
XM_015126746.2 | XP_014982232.1 | 552 | 152 | |||
XM_015126747.2 | XP_014982233.2 | 388 | 152 | |||
XM_015126748.2 | XP_014982234.2 | 380 | 152 | |||
GenBank mRNA | JU478221.1 | - | 4 - 5 | 435 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
