Assay Details
Gene Symbol: MRPS34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257615.1 | NP_001244544.1 | 378 | 109 | ||
GenBank mRNA | JU323144.1 | - | 378 | 109 | ||
JU470787.1 | - | 377 | 109 | |||
JU470788.1 | - | 377 | 109 | |||
JV638746.1 | - | 384 | 109 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
